Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A1/S100B, Mouse, Clone: 4B3, Abnova™

Catalog No. 89111215 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-215 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-215 Supplier Abnova Corporation Supplier No. MAB3031
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native S100A1/S100B.

Specifications

Antigen S100A1/S100B
Applications Western Blot
Classification Monoclonal
Clone 4B3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native S100A1/S100B.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene Alias S100/S100-alpha/S100A
Gene Symbols S100A1
Host Species Mouse
Immunogen Native purified human brain S100A1/S100B.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6271/6285
Test Specificity This antibody is specific to S100 beta-beta and S100 alpha-beta.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.