Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A14, Mouse, Clone: 5, Abnova™

Catalog No. 89113899 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-899 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-899 Supplier Abnova Corporation Supplier No. MAB9941
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant S100A14.

S100A14 is a member of a subfamily of proteins related by Ca(2+)-binding motifs to the EF-hand Ca(2+)-binding protein superfamily.[supplied by OMIM

Specifications

Antigen S100A14
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant S100A14.
Dilution The optimal working dilution should be determined by the end user.
Formulation In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300)
Gene S100A14
Gene Accession No. NM_020672.1
Gene Alias BCMP84/S100A15
Gene Symbols S100A14
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human S100A14.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57402
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.