Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A14, Mouse, Clone: 78es, Abnova™

Catalog No. 89113632 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-632 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-632 Supplier Abnova Corporation Supplier No. MAB8875
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant S100A14.

S100A14 is a member of a subfamily of proteins related by Ca(2+)-binding motifs to the EF-hand Ca(2+)-binding protein superfamily.[supplied by OMIM

Specifications

Antigen S100A14
Applications ELISA
Classification Monoclonal
Clone 78es
Concentration 1 ug/uL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant S100A14.
Dilution The optimal working dilution should be determined by the end user.
Formulation In citrate-Tris-HCl, pH7.0 (0.02% Proclin 300)
Gene S100A14
Gene Alias BCMP84/S100A15
Gene Symbols S100A14
Host Species Mouse
Immunogen Recombinant protein corresponding to full length of human S100A14.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57402
Test Specificity This antibody recognizes human protein S100A14.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.