Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A14, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107181 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-181 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-181 Supplier Abnova Corporation Supplier No. H00057402A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant S100A14.

S100A14 is a member of a subfamily of proteins related by Ca(2+)-binding motifs to the EF-hand Ca(2+)-binding protein superfamily.[supplied by OMIM

Sequence: MGQCRSANAEDAQEFSDVERAIETLIKNFHQYSVEGGKETLTPSELRDLVTQQLPHLMPSNCGLEEKIANLGSCNDSKLEFRSFWELIGEAAKSVKLERPVRGH

Specifications

Antigen S100A14
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant S100A14.
Formulation 50% glycerol
Gene S100A14
Gene Accession No. NM_020672
Gene Alias BCMP84/S100A15
Gene Symbols S100A14
Host Species Mouse
Immunogen S100A14 (NP_065723, 1 a.a. ∽ 104 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57402
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.