Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UCHL1, Mouse, Clone: YK-3E3, Abnova™

Catalog No. 89108770 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-770 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-770 Supplier Abnova Corporation Supplier No. MAB10579

Mouse monoclonal antibody raised against full-length recombinant S100A3.

The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein has the highest content of cysteines of all S100 proteins, has a high affinity for Zinc, and is highly expressed in human hair cuticle. The precise function of this protein is unknown. [provided by RefSeq

Specifications

Antigen UCHL1
Applications ELISA, Western Blot
Classification Monoclonal
Clone YK-3E3
Concentration 1.0 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant S100A3.
Dilution Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user.
Formulation In 20mM HEPES, pH 7.5, 150mM NaCl (50% glycerol, 0.1mg/mL BSA)
Gene S100A3
Gene Alias S100E
Gene Symbols S100A3
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human UCHL1.
Purification Method Protein A-Sepharose chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6274
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.