Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A7, Mouse, Clone: 3E11, Abnova™

Catalog No. 89105390 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-390 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-390 Supplier Abnova Corporation Supplier No. H00006278M07A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant S100A7.

The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein differs from the other S100 proteins of known structure in its lack of calcium binding ability in one EF-hand at the N-terminus. This protein is markedly over-expressed in the skin lesions of psoriatic patients, but is excluded as a candidate gene for familial psoriasis susceptibility. The exact function of this protein is not known. [provided by RefSeq

Sequence: MSNTQAERSIIGMIDMFHKYTRRDDKIDKPSLLTMMKENFPNFLSACDKKGTNYLADVFEKKDKNEDKKIDFSEFLSLLGDIATDYHKQSHGAAPCSGGSQ

Specifications

Antigen S100A7
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3E11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant S100A7.
Formulation ascites with no preservative
Gene S100A7
Gene Accession No. BC034687.1
Gene Alias PSOR1/S100A7c
Gene Symbols S100A7
Host Species Mouse
Immunogen S100A7 (AAH34687.1,1 a.a. ∽ 101 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6278
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.