Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-S100A9, Clone: 6B4,

Catalog No. MABF854MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF854MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF854MI Supplier MilliporeSigma™ Supplier No. MABF854
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects S100A9 clone: 6B4 in Human samples, and it is validated for ELISA, Western Blotting

Protein S100-A9 (UniProt P06702; also known as Calgranulin-B, Calprotectin L1H subunit, Leukocyte L1 complex heavy chain, Migration inhibitory factor-related protein 14, MRP-14, p14, S100 calcium-binding protein A9, S100A9) is encoded by the S100A9 (also known as 60B8AG, CAGB, CFAG, CGLB, L1AG, LIAG, MAC387, MRP14) gene (Gene ID 6280) in human. S100A9 is one of the 21 members of the S100 family of proteins. S100A8 (calgranulin-A), S100A9 (calgranulin-B) and the heterdimer S100A8/S100A9 (calprotectin) exert their immunological and inflammatory action via binding to TLR-4, RAGE and some other putative receptors. S100A8, calprotectin, and S100A9 are chemoattractants for neutrophils, and S100A9 is known to induce the expression and activation of beta2-integrins. S100A9 is also shown to stimulate neutrophil degranulation and phagocytosis, while both S100A8 and S100A9 can induce the secretion of inflammatory cytokines from human monocytes. In addition, S100A9 is shown to potentiate both fMLP- and GM-CSF-stimulated IL-8 production from neutrophils, although S100A9 on its own cannot induce neutrophil cytokine production in the absence of fMLP and GM-CSF.
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100A9
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6B4
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing PBS without preservatives.
Gene Accession No. P06702
Gene Symbols S100A9; CAGB; CFAG; MRP14
Host Species Mouse
Immunogen Recombinant protein corresponding to human S100A9.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_002956
Target Species Human
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.