Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100B, Mouse, Clone: SH-B1, Abnova™

Catalog No. 89111477 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-477 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-477 Supplier Abnova Corporation Supplier No. MAB3572
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native S100B.

Specifications

Antigen S100B
Applications Immunohistochemistry
Classification Monoclonal
Clone SH-B1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native S100B.
Dilution Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user.
Formulation In phosphate buffer, pH 7.3 (0.09% sodium azide)
Gene S100B
Gene Alias MGC128275
Gene Symbols S100B
Host Species Mouse
Immunogen Native purified S100B from bovine brain.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 525716
Test Specificity This antibody reacts with S100 beta, S-100 A (alpha/beta) and S-100 B (beta/beta), not reactive with S-100 A0 (alpha/alpha), Calmodulin, Parvalbumin and other Ca-binding proteins.
Target Species Bovine, Feline, Human, Pig, Rabbit, Rat
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.