Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S1PR3, Mouse, Clone: Mix, Abnova™

Catalog No. 89111587 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-587 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-587 Supplier Abnova Corporation Supplier No. MAB3773

Mouse monoclonal antibody raised against synthetic peptide of S1PR3.

This gene encodes a member of the EDG family of receptors, which are G protein-coupled receptors. This protein has been identified as a functional receptor for sphingosine 1-phosphate and likely contributes to the regulation of angiogenesis and vascular endothelial cell function. [provided by RefSeq

Specifications

Antigen S1PR3
Applications Western Blot
Classification Monoclonal
Clone Mix
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of S1PR3.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene S1PR3
Gene Alias EDG-3/EDG3/FLJ37523/FLJ93220/LPB3/MGC71696/S1P3
Gene Symbols S1PR3
Host Species Mouse
Immunogen A synthetic peptide corresponding to C-terminus of human S1PR3.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1903
Test Specificity Recognizes EDG-3 (CT). Does not recognize EDG-2, 4 and 5. Recognizes a band of approximately 45 KDa in human and rat spleen and lung tissue (5 ug protein sample) using standard procedures.
Target Species Human, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.