Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAG, Mouse, Clone: PDS-1, Abnova™

Catalog No. 89110927 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-927 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-927 Supplier Abnova Corporation Supplier No. MAB2318
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against SAG.

Specifications

Antigen SAG
Applications Immunofluorescence, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone PDS-1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against SAG.
Dilution Immunohistochemistry (Frozen sections) (1:100) The optimal working dilution should be determined by the end user.
Formulation In Tris-glycine, 150mM NaCl (0.05% sodium azide)
Gene SAG
Gene Symbols SAG
Host Species Mouse
Immunogen Pig SAG.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 397151
Test Specificity This antibody is specific to arrestin protein.
Target Species Bovine, Human, Pig
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.