Learn More
SAPS2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human SAPS2 protein.
Supplier: Abnova Corporation H00009701B01P
Description
Protein phosphatase regulatory subunits, such as SAPS2, modulate the activity of protein phosphatase catalytic subunits by restricting substrate specificity, recruiting substrates, and determining the intracellular localization of the holoenzyme. SAPS2 is a regulatory subunit for the protein phosphatase-6 catalytic subunit (PPP6C; MIM 300141) (Stefansson and Brautigan, 2006 [PubMed 16769727]).[supplied by OMIM
Sequence: MFWKFDLNTTSHVDKLLDKEHVTLQELMDEDDILQECKAQNQKLLDFLCRQQCMEELVSLITQDPPLDMEEKVRFKYPNTACELLTCDVPQISDRLGGDESLLSLLYDFLDHEPPLNPLLASFFSKTIGNLIARKTEQVITFLKKKDKFISLVLKHIGTSALMDLLLRLVSCVEPAGLRQDVLHWLNEEKVIQRLVELIHPSQDEDRQSNASQTLCDIVRLGRDQGSQLQEALEPDPLLTALESQDCVEQLLKNMFDGDRTESCLVSGTQVLLTLLETRRVGTEGLVDSFSQGLERSYAVSSSVLHGIEPRLKDFHQLLLNPPKKKAILTTIGVLEEPLGNARLHGARLMAALLHTNTPSINQELCRLNTMDLLLDLFFKYTWNNFLHFQVELCIAAILSHAAREERTEASGSESRVEPPHENGNRSLETPQPAASLPDNTMVTHLFQKCCLVQRILEAWEANDHTQAAGGMRRGNMGHLTRIANAVVQNLERGPVQTHISEVIRGLPADCRGRWESFVEETLTETNRRNTVDLAFSDYQIQQMTANFVDQFGFNDEEFADQDDNINAPFDRIAEINFNIDADEDSPSAALFEACCSDRIQPFDDDEDEDIWEDSDTRCAARVMARPRFGAPHASESCSKNGPERGGQDGKASLEAHRDAPGAGAPPAPGKKEAPPVEGDSEGAMWTAVFDEPANSTPTAPGVVRDVGSSVWAAGTSAPEEKGWAKFTDFQPFCCSESGPRCSSPVDTECSHAEGSRSQGPEKAFSPASPCAWNVCVTRKAPLLASDSSSSGGSHSEDGDQKAASAMDAVSRGPGREAPPLPTVARTEEAVGRVGCADSRLLSPACPAPKEVTAAPAVAVPPEATVAITTALSKAGPAIPTPAVSSALAVAVPLGPIMAVTAAPAMVATLGTVTKDGKTDAPPEGAALNGPVSpecifications
SAPS2 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human SAPS2 protein. | |
SAPS2 | |
KIAA0685/PP6R2/SAP190/dJ579N16.1 | |
Mouse | |
Affinity chromatography | |
RUO | |
9701 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_014678.3 | |
SAPS2 | |
SAPS2 (NP_055493.2,1 a.a. ∽ 932 a.a) full-length human protein. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only