Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SBF2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107396 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-396 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-396 Supplier Abnova Corporation Supplier No. H00081846A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant SBF2.

This gene encodes a pseudophosphatase and member of the myotubularin-related protein family. This gene maps within the CMT4B2 candidate region of chromosome 11p15 and mutations in this gene have been associated with Charcot-Marie-Tooth Disease, type 4B2. [provided by RefSeq

Sequence: MARLADYFIVVGYDHEKPGSGEGLGKIIQRFPQKDWDDTPFPQGIELFCQPGGWQLSRERKQPTFFVVVLTDIDSDRHYCSCLTFYEAEINLQGTKKEEI

Specifications

Antigen SBF2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant SBF2.
Formulation 50% glycerol
Gene SBF2
Gene Accession No. NM_030962
Gene Alias CMT4B2/DKFZp779B2327/FLJ22918/FLJ41627/KIAA1766/MTMR13
Gene Symbols SBF2
Host Species Mouse
Immunogen SBF2 (NP_112224, 1 a.a. ∽ 100 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81846
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.