Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCML4, Mouse, Clone: 2G12, Abnova™

Catalog No. 89107733 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-733 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-733 Supplier Abnova Corporation Supplier No. H00256380M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant SCML4.

Sequence: MPCQRTAWIGCDRQRTPPFHWREIKSRVLMTPLALSPPRSTPEPDLSSIPQDAATVPSLAAPQALTVCLYINKQANAGPYLERKKVQQLPEHFGPERPSAVLQQAVQACIDCAHQQKLVFSLVKQGYGGEMVSVSASFDGKQHLRSLPVVNSIGYVLRFLAKLCRSLLCDDLFSHQPFPRGCSASEKVQEKEEGRMESVKTVTTEEYLVNPVGMNRYSVDTSASTFNHRGSLHPSSSLYCKRQNSGDSHLGGGPAATAGGPRTSPMSSGGPSAPGLRPPASSPKRNTTSLEGNRCGNVMHASASH

Specifications

Antigen SCML4
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2G12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant SCML4.
Formulation PBS with no preservative; pH 7.4
Gene SCML4
Gene Accession No. NM_198081.2
Gene Alias FLJ36252/dJ47M23.1
Gene Symbols SCML4
Host Species Mouse
Immunogen SCML4 (NP_932347.1,1 a.a. ∽ 305 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A Purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 256380
Target Species Human, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.