Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scn2a1, Mouse, Clone: S69-3, Abnova™

Catalog No. 89113517 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-517 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-517 Supplier Abnova Corporation Supplier No. MAB8587

Mouse monoclonal antibody raised against partial recombinant Scn2a1.

Specifications

Antigen Scn2a1
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone S69-3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant Scn2a1.
Dilution Western Blot (1 ug/mL) Immunohistochemistry (0.1-1.0 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Scn2a1
Gene Alias NachII/Nav1.2/RII/RIIA/RNSCPIIR/SCN/SCN2A/Scn2a2/ScpII
Gene Symbols Scn2a1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1882-2005 of rat Scn2a1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24766
Test Specificity This antibody is specific to Scn2a1.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.