Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-Secretory Component, Clone: SC05/COMPO2, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2471380.2MG
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Secretory Component | |
Monoclonal | |
Unconjugated | |
10mM PBS - BSA free with No Preservative | |
Free Secretory Component (FSC), pIgA-Secretory Component Complex, pIgM-Secretory Component Complex, Secretory Component (SC), sIgA-Secretory Component Complex | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
The antibody reacts with a reduction-resistant epitope present in both free and SIgA bound Secretory Component. However, the exact epitope recognized by the antibody has not been mapped. | |
Store at 4C. | |
IgG1 κ |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence | |
SC05/COMPO2 | |
Flow Cytometry 0.5-1ug/million cells in 0.1ml, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunofluorescence 1-2ug/ml | |
P01833 | |
RIGR | |
Partially purified preparation of Secretory Component from human colostrum. | |
0.2 mg | |
Immunology | |
5284 | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction