Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SELE, Mouse, Clone: ELAM-1, Abnova™

Catalog No. 89112950 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-950 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-950 Supplier Abnova Corporation Supplier No. MAB6596
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native SELE.

The protein encoded by this gene is found in cytokine-stimulated endothelial cells and is thought to be responsible for the accumulation of blood leukocytes at sites of inflammation by mediating the adhesion of cells to the vascular lining. It exhibits structural features such as the presence of lectin- and EGF-like domains followed by short consensus repeat (SCR) domains that contain 6 conserved cysteine residues. These proteins are part of the selectin family of cell adhesion molecules. Adhesion molecules participate in the interaction between leukocytes and the endothelium and appear to be involved in the pathogenesis of atherosclerosis. [provided by RefSeq

Specifications

Antigen SELE
Applications ELISA
Classification Monoclonal
Clone ELAM-1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native SELE.
Dilution ELISA (1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (10mM sodium azide)
Gene SELE
Gene Alias CD62E/ELAM/ELAM1/ESEL/LECAM2
Gene Symbols SELE
Host Species Mouse
Immunogen Native purified SELE from human neutrophils.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6401
Test Specificity This antibody is specific to the human antigen E-selectin (ELAM-1, 115 KDa).
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.