Learn More
SELL, Mouse, Clone: DREG56, Abnova™
Mouse monoclonal antibody raised against native SELL.
Supplier: Abnova Corporation MAB8213
Description
SELL is a cell surface component that is a member of a family of adhesion/homing receptors which play important roles in leukocyte-endothelial cell interactions. The molecule is composed of multiple domains: one homologous to lectins, one to epidermal growth factor, and two to the consensus repeat units found in C3/C4 binding proteins. [provided by RefSeq
Specifications
SELL | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (2 ug/mL) Immunohistochemistry Immunoprecipitation Western Blot The optimal working dilution should be determined by the end user. | |
SELL | |
SELL | |
Native purified SELL from PMA-activated human peripheral blood leukocytes. | |
RUO | |
6402 | |
Human | |
Liquid |
ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
DREG56 | |
Mouse monoclonal antibody raised against native SELL. | |
In PBS, pH 7.4 (0.09% sodium azide) | |
CD62L/LAM-1/LAM1/LECAM1/LNHR/LSEL/LYAM1/Leu-8/Lyam-1/PLNHR/TQ1/hLHRc | |
Mouse | |
100 μg | |
Primary | |
This antibody recognizes SELL, a 65-76 KDa cell surface protein, expressed by neutrophils, monocytes, and subsets of T, B, and NK cells, that interacts with specific carbohydrates exposed on activated endothelial cells. | |
Store at 4°C. Do not freeze. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.