Learn More
SELL, Mouse, FITC, Clone: DREG56, Abnova™
Mouse monoclonal antibody raised against native SELL.
Supplier: Abnova Corporation MAB9801
Description
SELL is a cell surface component that is a member of a family of adhesion/homing receptors which play important roles in leukocyte-endothelial cell interactions. The molecule is composed of multiple domains: one homologous to lectins, one to epidermal growth factor, and two to the consensus repeat units found in C3/C4 binding proteins. [provided by RefSeq
Specifications
SELL | |
Monoclonal | |
FITC | |
Flow Cytometry (20 uL reagent/100 uL of whole blood or 106 cells) The optimal working dilution should be determined by the end user. | |
SELL | |
SELL | |
Native purified SELL from PMA-activated human peripheral blood leukocytes. | |
RUO | |
6402 | |
Human | |
Liquid |
Flow Cytometry | |
DREG56 | |
Mouse monoclonal antibody raised against native SELL. | |
In PBS, pH 7.4 (0.02% BSA, 0.09% sodium azide) | |
CD62L/LAM-1/LAM1/LECAM1/LNHR/LSEL/LYAM1/Leu-8/Lyam-1/PLNHR/TQ1/hLHRc | |
Mouse | |
100 Reactions | |
Primary | |
The mouse monoclonal antibody DREG56 recognizes SELL (CD62L / L-selectin), a 65-76 kDa cell surface protein, expressed by neutrophils, monocytes, and subsets of T, B, and NK cells, that interacts with specific carbohydrates exposed on activated endothelial cells. | |
Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze. | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.