Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SELL, Mouse, PE, Clone: LT-TD180, Abnova™

Catalog No. 89111833 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-833 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-833 Supplier Abnova Corporation Supplier No. MAB4452
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native SELL.

SELL is a cell surface component that is a member of a family of adhesion/homing receptors which play important roles in leukocyte-endothelial cell interactions. The molecule is composed of multiple domains: one homologous to lectins, one to epidermal growth factor, and two to the consensus repeat units found in C3/C4 binding proteins. [provided by RefSeq

Specifications

Antigen SELL
Applications Flow Cytometry
Classification Monoclonal
Clone LT-TD180
Conjugate PE
Description Mouse monoclonal antibody raised against native SELL.
Dilution Flow Cytometry (20 ul in human blood cells 100 ul in whole blood or 106 cells in a suspension) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene SELL
Gene Alias CD62L/LAM-1/LAM1/LECAM1/LNHR/LSEL/LYAM1/Leu-8/Lyam-1/PLNHR/TQ1/hLHRc
Gene Symbols SELL
Host Species Mouse
Immunogen Native purified SELL from peripheral blood leukocytes.
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6402
Test Specificity This antibody reacts with CD62L (L-selectin), a 74-95 KDa single chain type I glycoprotein expressed on most peripheral blood B lymphocytes, T lymphocytes, monocytes and granulocytes; it is also present on a subset of NK cells and certain hematopoietic malignant cells.
Target Species Human
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.