Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SELP, Mouse, APC, Clone: HI62P, Abnova™

Catalog No. 89111834 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-834 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-834 Supplier Abnova Corporation Supplier No. MAB4453

Mouse monoclonal antibody raised against native SELP.

This gene encodes a 140kDa protein that is stored in the alpha-granules of platelets and Weibel-Palade bodies of endothelial cells. This protein redistributes to the plasma membrane during platelet activation and degranulation and mediates the interaction of activated endothelial cells or platelets with leukocytes. The membrane protein is a calcium-dependent receptor that binds to sialylated forms of Lewis blood group carbohydrate antigens on neutrophils and monocytes. Alternative splice variants may occur but are not well documented. [provided by RefSeq

Specifications

Antigen SELP
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone HI62P
Conjugate APC
Description selectin P (granule membrane protein 140kDa, antigen CD62)
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene SELP
Gene Alias CD62, CD62P, FLJ45155, GMP140, GRMP, LECAM3, PADGEM, PSEL
Gene Symbols SELP
Host Species Mouse
Immunogen Native purified SELP from human platelets.
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6403
Target Species Human
Content And Storage Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.