Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEMA3A, Mouse, Clone: 5G9, Abnova™

Catalog No. 89106296 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-296 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-296 Supplier Abnova Corporation Supplier No. H00010371M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant SEMA3A.

This gene is a member of the semaphorin family and encodes a protein with an Ig-like C2-type (immunoglobulin-like) domain, a PSI domain and a Sema domain. This secreted protein can function as either a chemorepulsive agent, inhibiting axonal outgrowth, or as a chemoattractive agent, stimulating the growth of apical dendrites. In both cases, the protein is vital for normal neuronal pattern development. Increased expression of this protein is associated with schizophrenia and is seen in a variety of human tumor cell lines. Also, aberrant release of this protein is associated with the progression of Alzheimer's disease. [provided by RefSeq]

Sequence: HLEELLHKDDDGDGSKTKEMSNSMTPSQKVWYRDFMQLINHPNLNTMDEFCEQVWKRDRKQRRQRPGHTPGNSNKWKHLQENKKGRNRRTHEFERAPRS

Specifications

Antigen SEMA3A
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5G9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant SEMA3A.
Formulation ascites with no preservative
Gene SEMA3A
Gene Accession No. NM_006080
Gene Alias Hsema-I/Hsema-III/MGC133243/SEMA1/SEMAD/SEMAIII/SEMAL/SemD/coll-1
Gene Symbols SEMA3A
Host Species Mouse
Immunogen SEMA3A (NP_006071,672 a.a. ∽ 770 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10371
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.