Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sema4d, Mouse, Clone: SK-3, Abnova™

Catalog No. 89112946 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-946 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-946 Supplier Abnova Corporation Supplier No. MAB6583
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant Sema4d.

Specifications

Antigen Sema4d
Applications ELISA, Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone SK-3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant Sema4d.
Dilution ELISA (1-10 ug/mL) Immunoprecipitation (5 ug/mL) Flow Cyt (0.05 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.1% proclin, 2% Block Ace)
Gene Sema4d
Gene Alias CD100/Semacl2/Semaj/Semcl2/coll-4
Gene Symbols Sema4d
Host Species Mouse
Immunogen Recombinant protein corresponding to extracellular region of mouse Sema4d.
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20354
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.