Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sema7a, Mouse, Clone: SKK-7, Abnova™

Catalog No. 89112945 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-945 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-945 Supplier Abnova Corporation Supplier No. MAB6582
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of Sema7a.

Specifications

Antigen Sema7a
Applications ELISA, Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone SKK-7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of Sema7a.
Dilution ELISA (1.0-10 ug/mL) Flow Cyt (50 ng/mL) Immunoprecipitation (5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.1% proclin, 2% Block Ace)
Gene Sema7a
Gene Alias 2900057C09Rik/CDw108/H-Sema-L/M-Sema-L/Semal
Gene Symbols Sema7a
Host Species Mouse
Immunogen A synthetic peptide corresponding to extracellular domain of mouse Sema7a.
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20361
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.