Learn More
SEMG2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human SEMG2 protein.
Supplier: Abnova Corporation H00006407B01P
Description
The secreted protein encoded by this gene is involved in the formation of a gel matrix that encases ejaculated spermatozoa. Proteolysis by the prostate-specific antigen (PSA) breaks down the gel matrix and allows the spermatozoa to move more freely. The encoded protein is found in lesser abundance than a similar semenogelin protein. The genes encoding these two semenogelin proteins are found in a cluster on chromosome 20. [provided by RefSeq
Sequence: MKSIILFVLSLLLILEKQAAVMGQKGGSKGQLPSGSSQFPHGKKGQHYFGQKDQQHTKSKGSFSIQHTYHVDINDHDWTRKSQQYDLNALHKATKSKQHLGGSQQLLNYKQEGRDHDKSKGHFHMIVIHHKGGQAHHGTQNPSQDQGNSPSGKGLSSQCSNTEKRLWVHGLSKEQASASGAQKGRTQGGSQSSYVLQTEELVVNKQQRETKNSHQNKGHYQNVVDVREEHSSKLQTSLHPAHQDRLQHGPKDIFTTQDELLVYNKNQHQTKNLNQDQEHGRKAHKISYPSSRTEERQLHHGEKSVQKDVSKGSISIQTEEKIHGKSQNQVTIHSQDQEHGHKENKISYQSSSTEERHLNCGEKGIQKGVSKGSISIQTEEQIHGKSQNQVRIPSQAQEYGHKENKISYQSSSTEERRLNSGEKDVQKGVSKGSISIQTEEKIHGKSQNQVTIPSQDQEHGHKENKMSYQSSSTEERRLNYGGKSTQKDVSQSSISFQIEKLVEGKSQIQTPNPNQDQWSGQNAKGKSGQSADSKQDLLSHEQKGRYKQESSESHNIVITEHEVAQDDHLTQQYNEDRNPISTSpecifications
SEMG2 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human SEMG2 protein. | |
SEMG2 | |
SGII | |
Mouse | |
Affinity chromatography | |
RUO | |
6407 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC070306.1 | |
SEMG2 | |
SEMG2 (AAH70306.1,1 a.a. ∽ 582 a.a) full-length human protein. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only