Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-Ser/Thr-protein kinase Nek2, Clone: 5D2.1,

Catalog No. MABE974MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE974MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE974MI Supplier MilliporeSigma™ Supplier No. MABE974
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Ser/Thr-protein kinase Nek2 clone: 5D2.1 in Human, Mouse samples, and it is validated for Immunocytochemistry, Western Blotting

Serine/threonine-protein kinase Nek2 (EC 2.7.11.1; UniProt P51955; also known as HSPK 21, Never in mitosis A-related kinase 2, NimA-like protein kinase 1, NimA-related protein kinase 2, Protein phosphatase 1 regulatory subunit 111) is encoded by the NEK2 (also known as NEK2A, NLK1, PPP1R111, RP67) gene (Gene ID 4751) in human. Nek2 is a member of the never in mitosis A- (NIMA-) related family of serine/threonine kinases. Nek2 is involved in cell cycle regulation, gene expression, and the maintenance of centrosomal structure and function. Cellular Nek2 levels are tightly regulated throughout the cell cycle with noncentrosomal Nek2 expression peaks from S to G2 phase, while cytoplasmic Nek2 is highest in G1, and nuclear Nek2 is highest during S and G2 phase when it is needed for centrosomal function regulation. During interphase, centrosomes are held together by a proteinaceous linker that connects the the mother and daughter centriole. This linker is disassembled at the onset of mitosis in a process known as centrosome disjunction. The mammalian sterile 20-like kinase 2 (Mst2) and the scaffold protein Salvador (hSav1) are shown to directly interact with Nek2A and regulate its ability to localize to centrosomes, where Nek2A mediating linker dissociation by phosphorylating the centrosomal linker proteins C-Nap1 and rootletin. In addition, Nek2 is also reported to regulate gene expression by phosphorylating the splicing factor SRSF1. Nek2 expression is found upregulated in a wide variety of neoplasms including diffuse large B cell lymphoma and multiple myeloma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ser/Thr-protein kinase Nek2
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 5D2.1
Formulation Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P51955
Gene Symbols NEK2; NEK2A; NLK1
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to the N-terminal of human Ser/Thr-protein kinase Nek2.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_002488
Target Species Human, Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.