Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Mouse anti-Serotonin receptor 2A/HTR2A, Clone: 9B11.1,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABN1595
Description
Specifically detects Serotonin receptor 2A/HTR2A clone: 9B11.1 in Human, Mouse, Rat samples, and it is validated for Immunohistochemistry, Western Blotting
5-hydroxytryptamine receptor 2A (UniProt P28223; also known as 5-HT-2, 5-HT-2A, 5-HT2A, Serotonin receptor 2A) is encoded by the HTR2A (also known as HTR2) gene (Gene ID 3356) in human. Hydroxytryptamine (5-HT) is synthesized in enterochromaffin cells in the intestine and in serotonergic nerve terminals. In the periphery, 5-HT mediates gastrointestinal motility, platelet aggregation, and contraction of blood vessels. Many functions of the central nervous system are influenced by 5-HT, including sleep, motor activity, sensory perception, arousal and appetite. A family of 12 GPCRs and one ion channel mediate the biological effects of 5-HT. 5-HT2A receptor, which couples to Gq/11 to increase intracellular calcium, is widely expressed at central and peripheral sites of 5-HT action and contributes to many of the physiological effects of 5-HT. The hallucinogenic activity of LSD is mediated in part by its action as a partial to full agonist of 5-HT2A receptor, and the activity of atypical antipsychotics such as clozapine appears to be mediated in part by antagonism against 5-HT2A receptor. Human 5-HT2A receptor contains four extracellular domains (a.a. 1-75, 133-148, 216-233, 347-362), seven transmembrane segments (a.a. 76-99, 111-132, 149-171, 192-215, 234-254, 325-346, 363-384), and four cytoplasmic domains (a.a. 100-110, 172-191, 255-324, 385-471).
Specifications
Serotonin receptor 2A/HTR2A | |
Monoclonal | |
Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
HTR2A; HTR2 | |
GST-tagged recombinant fragment corresponding to the third cytoplasmic domain of human Serotonin receptor 2A/HTR2A. | |
100 μg | |
Neuroscience | |
NP_000612 | |
Stable for one year at 2°C to 8°C from date of receipt. | |
IgG2a κ |
Immunohistochemistry, Western Blot | |
9B11.1 | |
P28223 | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction