Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINA1 Mouse anti-Human, Monoclonal Antibody, Abnova™

Catalog No. 89112723 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-723 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-723 Supplier Abnova Corporation Supplier No. MAB6143
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native SERPINA1.

The protein encoded by this gene is secreted and is a serine protease inhibitor whose targets include elastase, plasmin, thrombin, trypsin, chymotrypsin, and plasminogen activator. Defects in this gene can cause emphysema or liver disease. Several transcript variants encoding the same protein have been found for this gene. [provided by RefSeq

Specifications

Antigen SERPINA1
Applications ELISA, Immunofluorescence
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native SERPINA1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Gene SERPINA1
Gene Alias A1A/A1AT/AAT/MGC23330/MGC9222/PI/PI1/PRO2275
Gene Symbols SERPINA1
Host Species Mouse
Immunogen Native purified SERPINA1 from human serum.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5265
Test Specificity These antibodies specifically recognize alpha-1-antichymotrypsin. They do not cross-react with alpha-1-antitrypsin.
Target Species Human
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.