Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINA12, Mouse, Clone: 1C4, Abnova™

Catalog No. 89109269 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-269 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-269 Supplier Abnova Corporation Supplier No. MAB1106
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant SERPINA12.

Specifications

Antigen SERPINA12
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1C4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant SERPINA12.
Dilution Western Blot (1:1000-1:3000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene SERPINA12
Gene Accession No. NM_173850
Gene Alias OL-64
Gene Symbols SERPINA12
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 21-414 of human SERPINA12.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 145264
Target Species Human, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.