Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-SERPINB5, Clone: 5C6.2,

Catalog No. MABC603MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC603MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC603MI Supplier MilliporeSigma™ Supplier No. MABC603
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects SERPINB5 clone: 5C6.2 in Human samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting

Serpin B5 (UniProt P36952; also known as Maspin, Peptidase inhibitor 5, PI-5) is encoded by the SERPINB5 (also known as PI5) gene (Gene ID 5268) in human. Serpin B5/Maspin is an evolutionarily conserved serine protease inhibitor that induces apoptosis and inhibits the motility of breast cancer cells. DNA hypermethylation and histone deacetylation lead to silencing of the SERPINB5 gene in human breast cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERPINB5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 5C6.2
Formulation Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P36952
Gene Symbols SERPINB5; PI5
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to the C-terminal of human SERPINB5.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_002630
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.