Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINB5,, Mouse, Clone: AT2N6, Abnova™

Catalog No. 89113392 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-392 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-392 Supplier Abnova Corporation Supplier No. MAB8227
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant SERPINB5.

Specifications

Antigen SERPINB5,
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone AT2N6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant SERPINB5.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene SERPINB5
Gene Alias PI5/maspin
Gene Symbols SERPINB5
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human SERPINB5.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5268
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.