Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINE1, Mouse, Clone: 1D5, Abnova™

Catalog No. 89108577 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-577 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-577 Supplier Abnova Corporation Supplier No. MAB10390
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant SERPINE1.

This gene encodes a member of the serine proteinase inhibitor (serpin) superfamily. This member is the principal inhibitor of tissue plasminogen activator (tPA) and urokinase (uPA), and hence is an inhibitor of fibrinolysis. Defects in this gene are the cause of plasminogen activator inhibitor-1 deficiency (PAI-1 deficiency), and high concentrations of the gene product are associated with thrombophilia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen SERPINE1
Applications ELISA, Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 1D5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant SERPINE1.
Dilution ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.5% protein stabilizer, 0.05% sodium azide)
Gene SERPINE1
Gene Alias PAI/PAI-1/PAI1/PLANH1
Gene Symbols SERPINE1
Host Species Mouse
Immunogen Recombinant protein corresponding to human SERPINE1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5054
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.