Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpine1, Mouse, Clone: H4B3, Abnova™

Catalog No. 89111604 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-604 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-604 Supplier Abnova Corporation Supplier No. MAB3806
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant Serpine1.

Specifications

Antigen Serpine1
Applications ELISA, Western Blot
Classification Monoclonal
Clone H4B3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant Serpine1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 50mM sodium phosphate buffer, 100mM NaCl, pH 7.4
Gene Serpine1
Gene Alias PAI-1/PAI1/Planh1
Gene Symbols Serpine1
Host Species Mouse
Immunogen Recombinant protein corresponding to mouse Serpine1.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18787
Test Specificity This IgG purified monoclonal antibody is inhibitory and specific to Mouse PAI-1.
Target Species Mouse
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.