Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpine1, Mouse, Clone: MA32K3, Abnova™

Catalog No. 89111602 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-602 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-602 Supplier Abnova Corporation Supplier No. MAB3804

Mouse monoclonal antibody raised against recombinant Serpine1.

Specifications

Antigen Serpine1
Applications ELISA
Classification Monoclonal
Clone MA32K3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant Serpine1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 50mM sodium phosphate buffer, 100mM NaCl, 1mM EDTA, pH 6.6
Gene Serpine1
Gene Alias PAI1A/Pai1/Pai1aa/Planh/RATPAI1A
Gene Symbols Serpine1
Host Species Mouse
Immunogen Recombinant protein corresponding to rat Serpine1.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24617
Test Specificity This IgG purified monoclonal antibody is specific to Rat PAI-1. It is non-inhibitory.
Target Species Rat
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.