Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINF1, Mouse, Clone: 14K, Abnova™

Catalog No. 89110586 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-586 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-586 Supplier Abnova Corporation Supplier No. MAB1712
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of SERPINF1.

The PEDF gene is a member of the serpin gene family. Serpins are a group of serine protease inhibitors, some of which have also been reported to exhibit neurotrophic activity.[supplied by OMIM

Specifications

Antigen SERPINF1
Applications Western Blot
Classification Monoclonal
Clone 14K
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of SERPINF1.
Dilution Western Blot (5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.1% BSA, 0.1% proclin)
Gene SERPINF1
Gene Alias EPC-1/PEDF/PIG35
Gene Symbols SERPINF1
Host Species Mouse
Immunogen A synthetic peptide (conjugated with KLH) corresponding to human SERPINF1.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5176
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.