Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINF1/SERPINF2, Mouse, Clone: 710-1E-94, Abnova™

Catalog No. 89111702 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-702 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-702 Supplier Abnova Corporation Supplier No. MAB4118
Add to Cart
Add to Cart
Request a Quote

Mouse monoclonal antibody raised against SERPINF1/SERPINF2.

Specifications

Antigen SERPINF1/SERPINF2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 710-1E-94
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against SERPINF1/SERPINF2.
Dilution Western Blot (reduced and non-reduced) (1-5 ug/mL) ELISA (1:50000) The optimal working dilution should be determined by the end user.
Formulation In 50mM sodium phosphate buffer, 100mM NaCl, 1mM EDTA, pH 6.6
Gene Alias EPC-1/PEDF/PIG35
Gene Symbols SERPINF1
Host Species Mouse
Immunogen Human SERPINF1/SERPINF2.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5176/5345
Test Specificity human Alpha 2 Antiplasmin.
Target Species Human
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.