Learn More
SETDB1, Mouse, Clone: 4E8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant SETDB1.
Supplier: Abnova Corporation H00009869M22A
Description
This gene encodes a histone methyltransferase. The encoded enzyme catalyzes the reaction of S-adenosyl-L-methionine and histone L-lysine to produce S-adenosyl-L-homocysteine and histone N(6)-methyl-L-lysine. The encoded protein likely functions in transcriptional repression. Alternatively spliced transcript variants have been described
Sequence: SSLPGCIGLDAATATVESEEIAELQQAVVEELGISMEELRHFIDEELEKMDCVQQRKKQLAELETWVIQKESEVAHVDQLFDDASRAVTNCESLVKDFYSpecifications
SETDB1 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
NM_012432 | |
SETDB1 | |
SETDB1 (NP_036564,2 a.a. ∽ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
9869 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Western Blot | |
4E8 | |
Mouse monoclonal antibody raised against a partial recombinant SETDB1. | |
SETDB1 | |
ESET/KG1T/KIAA0067/KMT1E | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.