Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SFRS12 (B01P), Mouse anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89107632 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-632 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-632 Supplier Abnova Corporation Supplier No. H00140890B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human SFRS12 protein.

SFRS12 belongs to the superfamily of serine/arginine-rich (SR) splicing factors. It modulates splice site selection by regulating the activities of other SR proteins (Barnard et al., 2002 [PubMed 11991645]).[supplied by OMIM

Sequence: MTSLMPGAGLLPIPTPNPLTTLGVSLSSLGAIPAAALDPNIATLGEIPQPPLMGNVDPSKIDEIRRTVYVGNLNSQTTTADQLLEFFKQVGEVKFVRMAGDETQPTRFAFVEFADQNSVPRALAFNGVMFGDRPLKINHSNNAIVKPPEMTPQAAAKELEEVMKRVREAQSFISAAIEPESGKSNERKGGRSRSHTRSKSRSSSKSHSRRKRSQSKHRSRSHNRSRSRQKDRRRSKSPHKKRSKSRERRKSRSRSHSRDKRKDTREKIKEKERVKEKDREKEREREKEREKEKERGKNKDRDKEREKDREKDKEKDREREREKEHEKDRDKEKEKEQDKEKEREKDRSKEIDEKRKKDKKSRTPPRSYNASRRSRSSSRERRRRRSRSSSRSPRTSKTIKRKSSRSPSPRSRNKKDKKREKERDHISERRERERSTSMRKSSNDRDGKEKLEKNSTSLKEKEHNKEPDSSVSKEVDDKDAPRTEENKIQHNGNCQLNEENLSTKTEAV

Specifications

Antigen SFRS12
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human SFRS12 protein.
Formulation PBS with no preservative; pH 7.4
Gene SFRS12
Gene Accession No. NM_139168.1
Gene Alias DKFZp564B176/MGC133045/SRrp508/SRrp86
Gene Symbols SFRS12
Host Species Mouse
Immunogen SFRS12 (NP_631907.1,1 a.a. ∽ 508 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140890
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.