Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SH2D3C, Mouse, Clone: 3B2, Abnova™

Catalog No. 89106203 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-203 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-203 Supplier Abnova Corporation Supplier No. H00010044M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant SH2D3C.

Sequence: MTAVGRRCPALGSRGAAGEPEAGSDYVKFSKEKYILDSSPEKLHKELEEELKLSSTDLRSHAWYHGRIPREVSETLVQRNGDFLIRDSLTSLGDYVLTCRWRNQALHFKI

Specifications

Antigen SH2D3C
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3B2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant SH2D3C.
Formulation ascites with no preservative
Gene SH2D3C
Gene Accession No. NM_005489
Gene Alias CHAT/FLJ39664/NSP3/PRO34088
Gene Symbols SH2D3C
Host Species Mouse
Immunogen SH2D3C (NP_005480,1 a.a. ∽ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10044
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.