Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SH3GL2, Mouse, Clone: 2C4, Abnova™

Catalog No. 89105443 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-443 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-443 Supplier Abnova Corporation Supplier No. H00006456M10A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant SH3GL2.

Sequence: MSVAGLKKQFHKATQKVSEKVGGAEGTKLDDDFKEMERKVDVTSRAVMEIMTKTIEYLQPNPASRAKLSMINTMSKIRGQGKGPGYPQAEALLAEAMLKFGRELGDDCNFGPALGEVGEAMRELSEVKDSLDIEVKQNFIDPLQNLHDKDLREIQHHLKKLEGRRLDFDYKKERQGKIPDEELHQALEKFDESKEIAESSMFNLLEMDIEQVSQLSALVQAQLEYHKQAVQILQQVTVRLEERIRQASSQPRREYQPKPRMSLEFPTGDSTQPNGGLSH

Specifications

Antigen SH3GL2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2C4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant SH3GL2.
Formulation ascites with no preservative
Gene SH3GL2
Gene Accession No. BC032825
Gene Alias CNSA2/EEN-B1/FLJ20276/FLJ25015/SH3D2A/SH3P4
Gene Symbols SH3GL2
Host Species Mouse
Immunogen SH3GL2 (AAH32825,1 a.a. ∽ 279 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6456
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.