Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Shank1, Mouse, Clone: S22-21, Abnova™

Catalog No. 89113214 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-214 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-214 Supplier Abnova Corporation Supplier No. MAB7892
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant Shank1.

Specifications

Antigen Shank1
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone S22-21
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant Shank1.
Dilution Immunocytochemistry (1:100) Immunofluorescence (1:100) Immunohistochemistry (1:1000) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Shank1
Gene Alias Shank1a/Spank1/Sstrip
Gene Symbols Shank1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 469-691 of rat Shank1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 78957
Test Specificity Detects ∼190-220 kDa (alternative splice variants). No cross-reactivity against Shank2 or Shank3.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.