Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Shank2, Mouse, Clone: S23B-49, Abnova™

Catalog No. 89113212 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-212 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-212 Supplier Abnova Corporation Supplier No. MAB7890
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant Shank2.

Specifications

Antigen Shank2
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone S23B-49
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant Shank2.
Dilution Western Blot (1-10 ug/mL) Immunohistochemistry (0.1-1.0 ug/mL) Immunocytochemistry (0.1-1.0 ug/mL) Immunofluorescence (1-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Shank2
Gene Alias CortBP1/ProSAP1/Spank-3
Gene Symbols Shank2
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 84-309 of rat Shank2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171093
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.