Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHANK2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106539 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-539 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-539 Supplier Abnova Corporation Supplier No. H00022941A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant SHANK2.

This gene encodes a protein that is a member of the Shank family of synaptic proteins that may function as molecular scaffolds in the postsynaptic density (PSD). Shank proteins contain multiple domains for protein-protein interaction, including ankyrin repeats, an SH3 domain, a PSD-95/Dlg/ZO-1 domain, a sterile alpha motif domain, and a proline-rich region. This particular family member contains a PDZ domain, a consensus sequence for cortactin SH3 domain-binding peptides and a sterile alpha motif. The alternative splicing demonstrated in Shank genes has been suggested as a mechanism for regulating the molecular structure of Shank and the spectrum of Shank-interacting proteins in the PSDs of adult and developing brain. Two alternative splice variants, encoding distinct isoforms, are reported. Additional splice variants exist but their full-length nature has not been determined. [provided by RefSeq

Sequence: SILQQMNREKLAKPGEGLDSPMGAKSASLAPRSPEIMSTISGTRSTTVTFTVRPGTSQPITLQSRPPDYESRTSGTRRAPSPVVSPTEMNK

Specifications

Antigen SHANK2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant SHANK2.
Formulation 50% glycerol
Gene SHANK2
Gene Accession No. NM_012309
Gene Alias CORTBP1/CTTNBP1/ProSAP1/SHANK/SPANK-3
Gene Symbols SHANK2
Host Species Mouse
Immunogen SHANK2 (NP_036441, 1034 a.a. ∽ 1124 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22941
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.