Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-Shank3, Clone: 16E2.3,

Catalog No. MABN790MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN790MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN790MI Supplier MilliporeSigma™ Supplier No. MABN790
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Shank3 clone: 16E2.3 in Mouse samples, and it is validated for Western Blotting

SH3 and multiple ankyrin repeat domains protein 3 (UniProt Q9BYB0; also known as Proline rich synapse associated protein 2, ProSAP2, Shank postsynaptic density protein, Shank3) is encoded by the SHANK3 (also known as DEL22q13.3, PROSAP2, PSAP2, SCZD15, SPANK-2) gene (Gene ID 85358) in human. Shank3/ProSAP2 belongs to the ProSAP/Shank family of scaffolding proteins, which are important dendritic spines postsynaptic density (PSD) complex components that play an important role in linking membrane glutamate receptors to intracellular signaling proteins and actin cytoskeleton. Shank proteins interact with adaptor proteins and assembles glutamate receptors at the postsynaptic membrane via multiple domains, including ankyrin repeat, proline-rich, SH3, PSD95/disc-large/zona occludens-1 (PDZ), and SAM domains. Shanks, together with the adaptor protein Homer and the endocytic vesicle-associated protein dynamin-3, are required for proper localization of the endocytic zone in close proximity to the PSD. Shank-3 induces formation of functional dendritic spines in aspiny cerebellar neurons, whereas Shank-3 knockdown results in reduced spine density and size in cultured hippocampal neurons. SHANK3 gene mutations are known causes of mental retardation and autism spectrum disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Shank3
Applications Western Blot
Classification Monoclonal
Clone 16E2.3
Formulation Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q9BYB0
Gene Symbols SHANK3; KIAA1650; PROSAP2; PSAP2
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to the C-terminal half of human Shank3.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_277052
Target Species Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.