Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHISA5 (B02P), Mouse anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89106871 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-871 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-871 Supplier Abnova Corporation Supplier No. H00051246B02P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human SHISA5 protein.

Sequence: MGFGATLAVGLTIFVLSVVTIIICFTCSCCCLYKTCRRPRPVVTTTTSTTVVHAPYPQPPSVPPSYPGPSYQGYHTMPPQPGMPAAPYPMQYPPPYPAQPMGPPAYHETLAGGAAAPYPASQPPYNPAYMDAPKAAL

Specifications

Antigen SHISA5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human SHISA5 protein.
Formulation PBS with no preservative; pH 7.4
Gene SHISA5
Gene Accession No. BC001463
Gene Alias SCOTIN/hShisa5
Gene Symbols SHISA5
Host Species Mouse
Immunogen SHISA5 (AAH01463,1 a.a. ∽ 137 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51246
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.