Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIAH1/SIAH2, Mouse, Clone: 8G7H12, Abnova™

Catalog No. 89135162 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-162 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-162 Supplier Abnova Corporation Supplier No. MAB2299
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of SIAH1/SIAH2.

Specifications

Antigen SIAH1/SIAH2
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 8G7H12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of SIAH1/SIAH2.
Dilution Western Blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In tissue culture supernatant (0.09% sodium azide)
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 280-331 of drosophila SIAH1/SIAH2.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody is specific to human SIAH 1 and 2 and Drosophila SINA in proliferating cells and stem cells. This antibody does not detect Drosophila SINAH or normal, nondividing cells.
Target Species Drosophila, Human, Pig, Rat, Zebrafish
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.