Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sialoganglioside, Mouse, Clone: eBio3G11, Abnova™

Catalog No. 89134837 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-837 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-837 Supplier Abnova Corporation Supplier No. MAB11083

Mouse monoclonal antibody raised against Sialoganglioside Antigen.

Specifications

Antigen Sialoganglioside
Applications Flow Cytometry
Classification Monoclonal
Clone eBio3G11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Sialoganglioside Antigen.
Dilution The optimal working dilution should be determined by the end user.
Formulation In aqueous buffer (0.09% sodium azide)
Host Species Mouse
Immunogen Mouse and rat sialoganglioside antigen.
Purification Method Affinity purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store at 4°C.
Form Liquid
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.