Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sialosyl-Tn, Mouse, Clone: STn 218, Abnova™

Catalog No. 89136148 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-148 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-148 Supplier Abnova Corporation Supplier No. MAB6199
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Sialosyl-Tn antigen.

Specifications

Antigen Sialosyl-Tn
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone STn 218
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Sialosyl-Tn antigen.
Dilution Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (1% BSA, 0.05% sodium azide)
Host Species Mouse
Immunogen Ovine submaxillary mucin (OSM).
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody stains colon carcinomas and ovarian mucinous carcinomas producing apical cytoplasmic, apical membrane or luminal staining patterns. It is negative with majority of adult tissue sections.
Target Species Human, Sheep
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.