Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sialyl LeA, Mouse, Clone: KM231, Abnova™

Catalog No. 89135434 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-434 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-434 Supplier Abnova Corporation Supplier No. MAB3342

Mouse monoclonal antibody raised against native Sialyl LeA.

Specifications

Antigen Sialyl LeA
Applications ELISA, Flow Cytometry, Immunohistochemistry, Radioimmune Assays (RIA)
Classification Monoclonal
Clone KM231
Conjugate Unconjugated
Formulation In PBS, pH 7.2 (0.09% sodium azide)
Host Species Mouse
Immunogen Native purified Sialyl LeA from human gastric cancer cells.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.