Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SILV, Mouse, Clone: NKI-beteb, Abnova™

Catalog No. 89109439 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-439 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-439 Supplier Abnova Corporation Supplier No. MAB11274
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against SILV.

Specifications

Antigen SILV
Applications Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone NKI-beteb
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against SILV.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:400) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene SILV
Gene Alias D12S53E/ME20/PMEL/PMEL17/SI/SIL/gp100
Gene Symbols SILV
Host Species Mouse
Immunogen Pigmented melanoma metastases extract isolated from lymph node.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6490
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.