Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIP1, Mouse, Clone: 2E17, Abnova™

Catalog No. 89111015 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-015 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-015 Supplier Abnova Corporation Supplier No. MAB2466
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant SIP1.

Specifications

Antigen SIP1
Applications ELISA, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2E17
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant SIP1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene SIP1
Gene Alias GEMIN2/SIP1-delta
Gene Symbols SIP1
Host Species Mouse
Immunogen Recombinant His fusion protein corresponding to human SIP1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8487
Test Specificity Detects a band of approximately 34 KDa.
Target Species Frog, Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.